accessibility menu, dialog, popup

UserName

Benzoyl peroxides

Organic compounds that contain two benzoyl groups bridged by a peroxide bond in the following formula: (C6H5CO)2O2.
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (1)
  • (1)
  • (3)
Grade 
  • (1)
Showing products from the Benzoyl peroxides category. View All Search Results

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)