missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric acid, 98+%

Catalog No. AC302511000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302511000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC302511000 Supplier Thermo Scientific Chemicals Supplier No. 302511000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17026-42-5 | C18H14O8 | 358.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Specific Rotation Condition +120° (20°C c=5,CH3OH)
Specific Rotation + 120
Solubility Information Solubility in water: insoluble
IUPAC Name 2,3-bis(benzoyloxy)butanedioic acid
Formula Weight 358.31
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-097-1

RUO – Research Use Only