missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene, 98%

Catalog No. AAA1228836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1228836 500 g
AAA1228814 25 g
AAA1228822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1228836 Supplier Thermo Scientific Chemicals Supplier No. A1228836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 132-65-0 | C12H8S | 184.256 g/mol

Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzothiophene is used to investigate the effect of sulfur compounds in gasoline range during the fluid catalytic cracking (FCC) process.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 132-65-0
Molecular Formula C12H8S
Molecular Weight (g/mol) 184.256
MDL Number MFCD00004969
InChI Key IYYZUPMFVPLQIF-UHFFFAOYSA-N
Synonym diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene
PubChem CID 3023
ChEBI CHEBI:23681
IUPAC Name dibenzothiophene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2

Specifications

Melting Point 97°C to 100°C
Boiling Point 332°C to 333°C
Flash Point 170°C (338°F)
Quantity 500 g
UN Number UN3077
Beilstein 121101
Solubility Information Soluble in water.
Formula Weight 184.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dibenzothiophene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H StatementH302 Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-072-9
RTECSNumber HQ3490550
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only