missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberenone 98.0+%, TCI America™

Catalog No. D179125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D179125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D179125G Supplier TCI America Supplier No. D179125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2222-33-5
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.244
MDL Number MFCD00003588
InChI Key SNVTZAIYUGUKNI-UHFFFAOYSA-N
Synonym dibenzosuberenone, 5-dibenzosuberenone, 5h-dibenzo a,d cyclohepten-5-one, 5h-dibenzo a,d 7 annulen-5-one, dibenzo a,d cyclohepten-5-one, 2,3:6,7-dibenzotropone, 2,3:6,7-dibenzosuberen-4-on-1, 5h-dibenzo a,d cycloheptenone
PubChem CID 16679
IUPAC Name dibenzo[1,2-a
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3C2=O

Specifications

Melting Point 89°C
Color White-Yellow
Boiling Point 210°C
Quantity 25 g
UN Number 3077
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzosuberenone
EINECSNumber (4)-1656
RTECSNumber HP1147500
TSCA Yes