missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo[a,e]cyclooctene 97.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 107°C |
| Color | White-Yellow |
| Boiling Point | 130°C |
| Quantity | 200 mg |
| Formula Weight | 204.27 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dibenzo[a,e]cyclooctene |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |