missing translation for 'onlineSavingsMsg'
Learn More

Dibenzenesulfonimide 98.0+%, TCI America™

Catalog No. D25405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25405G 5 g
D254025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25405G Supplier TCI America Supplier No. D25405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2618-96-4
Molecular Formula C12H11NO4S2
Molecular Weight (g/mol) 297.343
MDL Number MFCD00025019
InChI Key OVQABVAKPIYHIG-UHFFFAOYSA-N
Synonym dibenzenesulfonimide, benzenesulfonamide, n-phenylsulfonyl, dibenzenesulfonamide, bis phenylsulfonyl amine, n-phenylsulfonyl benzenesulfonamide, dibenzolsulfimid, dibenzolsulfimide, diphenylsulfonimide, benzenesulfonic acid imide, n-benzenesulfonyl benzenesulfonamide
PubChem CID 75671
IUPAC Name N-(benzenesulfonyl)benzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)NS(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Formula Weight 297.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzenesulfonimide
EINECSNumber (3)-3133
TSCA Yes