missing translation for 'onlineSavingsMsg'
Learn More

Diatrizoic Acid 98.0+%, TCI America™

Catalog No. D146225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D146225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D146225G Supplier TCI America Supplier No. D146225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117-96-4
Molecular Formula C11H9I3N2O4
Molecular Weight (g/mol) 613.92
MDL Number MFCD00069960
InChI Key YVPYQUNUQOZFHG-UHFFFAOYSA-N
Synonym diatrizoic acid, amidotrizoic acid, diatrizoate, amidotrizoate, urogranoic acid, urografin acid, odiston, urotrast, diat, diatriazoate
PubChem CID 2140
ChEBI CHEBI:53691
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid
SMILES CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I

Specifications

Color White
Quantity 25 g
Formula Weight 613.92
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diatrizoic Acid
RTECSNumber DG5950000
TSCA No