missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Diamond PosiStop Pipettors, Fixed Volume

Catalog No. 23763087
Change view
Click to view available options
Volume (Metric):
2.5 μL
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
500 μL
1000 μL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23763087 500 μL
23763080 2.5 μL
23763085 5 μL
23763077 10 μL
23763081 20 μL
23763083 25 μL
23763086 50 μL
23763078 100 μL
23763082 200 μL
23763084 250 μL
23763079 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 23763087 Supplier Globe Scientific Supplier No. 3362500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Discover the Diamond PosiStop LTS-compatible pipettors - engineered for precision, comfort, and durability. With a low-force plunger, shock-absorbing tip ejector, and robust PVDF construction, they deliver consistent performance and ergonomic ease. Backed by a 3-year warranty.

  • Fully Autoclavable and UV Resistant: Ensuring durability and adaptability in various laboratory environments.
  • 3-Year Warranty: Providing extended assurance of quality and performance.
  • Color-Coded Plungers: For quick and easy volume identification, streamlining your laboratory processes.
  • Low Plunger Force: Offering a virtually fatigue-free pipetting experience.
  • Magnetically Assisted Piston: Guarantees extremely high accuracy and consistency.
  • Ergonomically Designed and Lightweight: Enhances comfort during prolonged use.
  • Secure Locking Mechanism: Prevents accidental volume changes, ensuring reliable results.
  • Availability in Fixed Volume Models: Catering to diverse laboratory needs with specific requirements.

Specifications

Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette Tip 1000 μL
Material PVDF
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Pipette
Volume (Metric) 500 μL