missing translation for 'onlineSavingsMsg'
Learn More

Diallylamine 98.0+%, TCI America™

Numéro de catalogue. p-7126010
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D006925ML 25 mL
D0069500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D006925ML Fournisseur TCI America Code fournisseur D006925ML
Il en reste null

Identifiants chimiques

CAS 124-02-7
Molecular Formula C6H11N
Molecular Weight (g/mol) 97.161
MDL Number MFCD00008642
InChI Key DYUWTXWIYMHBQS-UHFFFAOYSA-N
Synonym diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine
PubChem CID 31279
IUPAC Name N-prop-2-enylprop-2-en-1-amine
SMILES C=CCNCC=C

Spécifications

Melting Point -88°C
Color Yellow
Boiling Point 112°C
Quantity 25 mL
UN Number 2359
Formula Weight 97.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallylamine
EINECSNumber (2)-0171
RTECSNumber UC6650000
TSCA Yes