missing translation for 'onlineSavingsMsg'
Learn More

Diallylamine 98.0+%, TCI America™

Catalog No. D0069500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0069500ML 500 mL
D006925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0069500ML Supplier TCI America Supplier No. D0069500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-02-7
Molecular Formula C6H11N
Molecular Weight (g/mol) 97.161
MDL Number MFCD00008642
InChI Key DYUWTXWIYMHBQS-UHFFFAOYSA-N
Synonym diallylamine, di-2-propenylamine, 2-propen-1-amine, n-2-propenyl, amine, diallyl, bis prop-2-en-1-yl amine, n-2-propenyl-2-propen-1-amine, unii-n18exb6v6p, n,n-diallylamine, ccris 4776, n-allylprop-2-en-1-amine
PubChem CID 31279
IUPAC Name N-prop-2-enylprop-2-en-1-amine
SMILES C=CCNCC=C

Specifications

Melting Point -88°C
Color Yellow
Boiling Point 112°C
Quantity 500 mL
UN Number 2359
Formula Weight 97.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallylamine
EINECSNumber (2)-0171
RTECSNumber UC6650000
TSCA Yes