missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallylphtalate 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 158°C |
| Quantity | 25 mL |
| UN Number | 3082 |
| Formula Weight | 246.26 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Diallyl Phthalate |
Sécurité et manipulation
| EINECSNumber | (3)-1325 |
| RTECSNumber | CZ4200000 |
| TSCA | Yes |