missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl Isophthalate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 163°C |
| Quantity | 500 g |
| Formula Weight | 246.26 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Diallyl Isophthalate |
Safety and Handling
| EINECSNumber | (3)-1317&(3)-1325&(3)-1329 |
| RTECSNumber | CZ4230000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |