missing translation for 'onlineSavingsMsg'
Learn More

Diallyl Disulfide 80.0+%, TCI America™

Catalog No. D007125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D007125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D007125G Supplier TCI America Supplier No. D007125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2179-57-9
Molecular Formula C6H10S2
Molecular Weight (g/mol) 146.27
MDL Number MFCD00008656
InChI Key PFRGXCVKLLPLIP-UHFFFAOYSA-N
Synonym diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
PubChem CID 16590
ChEBI CHEBI:4488
IUPAC Name 3-(prop-2-en-1-yldisulfanyl)prop-1-ene
SMILES C=CCSSCC=C

Specifications

Color Yellow
Quantity 25 g
UN Number 2810
Formula Weight 146.27
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Diallyl Disulfide
EINECSNumber (2)-0481
RTECSNumber BB1000000
TSCA Yes
Recommended Storage Refrigerator