missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Diaion™ HP-2MG
Description
Diaion™ HP-2MG is a methacrylic ester copolymer which is hydrophilic in nature. It is widely used for preconcentration and separation of traces of heavy metal ions. It is also used as adsorbent for polyphenols and surfactants.
Spécifications
Spécifications
| Pore Size | 170 Å |
| Quantity | 100 g |
| Surface Area (Metric) | ˜500 m2/g |
| Product Line | Diaion |
| Density | 1.09 g/mL at 25°C (true wet)(lit.) |
Diaion is a registered trademark of Mitsubishi Chemical Corp.