missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diacetyl-L-tartaric Anhydride 97.0+%, TCI America™

Catalog No. D191125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D191125G Supplier TCI America Supplier No. D191125G

Chemical Identifiers

CAS 6283-74-5
Molecular Formula C8H8O7
Molecular Weight (g/mol) 216.15
MDL Number MFCD00037918
InChI Key XAKITKDHDMPGPW-IOMOGOHMNA-N
Synonym 3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl diacetate, unii-d7xnf76o0k, +-diacetyl-l-tartaric anhydride, d7xnf76o0k, 2,5-furandione, 3,4-bis acetyloxy dihydro-, 3r,4r, 3r,4r-4-acetyloxy-2,5-dioxooxolan-3-yl acetate, di-o-acetyl-l-tartaric anhydride, o,o-diacetyl tartaric acid anhydride, acetic acid 4-acetoxy-2,5-dioxo-tetrahydro-furan-3-yl ester, 3r-trans-dihydro-2,5-dioxofuran-3,4-diyl diacetate
PubChem CID 513914
IUPAC Name (3R,4R)-4-(acetyloxy)-2,5-dioxooxolan-3-yl acetate
SMILES CC(=O)O[C@@H]1[C@@H](OC(C)=O)C(=O)OC1=O

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 216.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Diacetyl-L-tartaric Anhydride
TSCA No
Recommended Storage Freezer