missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyltin dichloride, 98%

Catalog No. AA3961904
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3961904 2 g
AA3961902 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3961904 Supplier Thermo Scientific Chemicals Supplier No. 03961904

missing translation for 'validMainframeMsg'

CAS: 19429-30-2 | C8H18Cl2Sn | 303.842 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19429-30-2
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00009854
InChI Key PEGCFRJASNUIPX-UHFFFAOYSA-L
Synonym di-tert-butyldichlorostannane, di-tert-butyltin dichloride, di-t-butyltin dichloride, ditert-butyl dichloro stannane, acmc-1bpuo, di-tert-butylzinndichlorid
PubChem CID 88053
IUPAC Name ditert-butyl(dichloro)stannane
SMILES CC(C)(C)[Sn](C(C)(C)C)(Cl)Cl

Specifications

Melting Point 42°C to 43°C
Color White
Boiling Point 66°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Linear Formula [(CH3)3C]2SnCl2
Quantity 2 g
UN Number UN2928
Sensitivity Moisture sensitive
Formula Weight 303.83
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Di-tert-butyltin dichloride
Show More Show Less
Hazard Category H301+H311+H331-H314
Hazard Statement GHS H Statement
H301-H311-H330-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 243-051-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only