missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl hydrazodicarboxylate, 98+%

Catalog No. AAL0028014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0028014 25 g
AAL0028006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0028014 Supplier Thermo Scientific Chemicals Supplier No. L0028014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16466-61-8 | C10H20N2O4 | 232.28 g/mol

Di-tert-butyl hydrazodicarboxylate, is used as starting reagent during the four-step synthesis of the aminobicyclopyrazolone hydrochloride. It was used as reagent during Pd-catalyzed amination reactions of halo-pyridine substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-tert-butyl hydrazodicarboxylate, is used as starting reagent during the four-step synthesis of the aminobicyclopyrazolone hydrochloride. It was used as reagent during Pd-catalyzed amination reactions of halo-pyridine substrates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 16466-61-8
Molecular Formula C10H20N2O4
Molecular Weight (g/mol) 232.28
MDL Number MFCD00015000
InChI Key TYSZETYVESRFNT-UHFFFAOYSA-N
Synonym di-tert-butyl hydrazodicarboxylate, di-tert-butyl hydrazine-1,2-dicarboxylate, di-tert-butyl hydrazodiformate, di-tert-butyl bicarbamate, n,n'-di-boc-hydrazine, n,n'-bis tert-butoxycarbonyl hydrazine, di-tert-butyl-1,2-hydrazodicarboxylate, 1,2-hydrazinedicarboxylic acid, bis 1,1-dimethylethyl ester, tert-butyl n-2-methylpropan-2-yl oxycarbonylamino carbamate, n'-tert-butoxy carbonyl tert-butoxy carbohydrazide
PubChem CID 85431
IUPAC Name tert-butyl N-[(2-methylpropan-2-yl)oxycarbonylamino]carbamate
SMILES CC(C)(C)OC(=O)NNC(=O)OC(C)(C)C

Specifications

Melting Point 123°C to 126°C
Quantity 25 g
Beilstein 1794023
Solubility Information Insoluble in water.
Formula Weight 232.28
Percent Purity ≥98%
Chemical Name or Material Di-tert-butyl hydrazodicarboxylate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-512-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only