missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Hydrogen Phosphate Dihydrate Granulated, Supelco

Numéro de catalogue. M1174175000
Modifier l'affichage
Click to view available options
Quantité:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
M1174175000 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M1174175000 Fournisseur Technidata Code fournisseur 1174175000

For Analysis EMSURE™

  • Experience the advantages of EMSURE™ Granulated Salts, specially designed for optimal handling and processing
  • Unlike powdered salts, our granulated formulation offers superior flow properties, ensuring consistency in your analytical applications
  • Our innovative dry granulation process keeps the salts free-flowing while preserving premium specifications, even during long-term storage

Key Benefits:

  • Exceptional Purity: Achieve reliable results with EMSURE™ analytical grade salts.
  • Chemical Stability: Ensure consistent outcomes with our stable formulation.
  • Enhanced Flowability: Enjoy easier handling, improved safety, and reduced waste during use.
  • Dust-Free Handling: Benefit from a clean, dust-free experience that enhances safety and usability.
  • Unchanged Specifications: Rest assured that product specifications remain consistent and optimized for analytical applications.

Identifiants chimiques

Forme physique Granules

Spécifications

Nom ou substance chimique di-Sodium Hydrogen Phosphate Dihydrate Granulated
CAS 10028-24-7
Quantité 5 kg
Pourcentage de pureté ≥99.5% (Acidimetric)