missing translation for 'onlineSavingsMsg'
Learn More

di-Sodium Hydrogen Phosphate, MilliporeSigma™

Catalog No. M1065860500
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065860500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065860500 Supplier Technidata Supplier No. 1.06586.0500
Only null left

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Melting Point 250°C (Decomposition)
pH 8.7 - 9.3
Quantity 500 g
Linear Formula HNa2O4P
Solubility Information 77g/L
Formula Weight 141.96
Loss on Drying ≤0.2%
Packaging Plastic Bottle
Density Bulk Density: 880 kg/m3
Assay ≥99.0%
Chemical Name or Material di-Sodium Hydrogen Phosphate
Show More Show Less
EINECSNumber 231-448-7,
RTECSNumber WC4500000
storageNote1 Store at +5°C to +30°C