missing translation for 'onlineSavingsMsg'
Learn More

(-)-Di-p-anisoyl-L-tartaric Acid 97.0+%, TCI America™

Catalog No. D349125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D349125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D349125G Supplier TCI America Supplier No. D349125G
Only null left

Chemical Identifiers

CAS 50583-51-2
Molecular Formula C20H18O10
Molecular Weight (g/mol) 418.354
MDL Number MFCD02682986
InChI Key KWWCVCFQHGKOMI-HZPDHXFCSA-N
Synonym (-)-Bis(4-methoxybenzoyl)-L-tartaric Acid
PubChem CID 11704418
IUPAC Name (2R,3R)-2,3-bis[(4-methoxybenzoyl)oxy]butanedioic acid
SMILES COC1=CC=C(C=C1)C(=O)OC(C(C(=O)O)OC(=O)C2=CC=C(C=C2)OC)C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 418.35
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Di-p-anisoyl-L-tartaric Acid