missing translation for 'onlineSavingsMsg'
Learn More

Di(N-succinimidyl) Suberate 98.0+%, TCI America™

Catalog No. D38955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38955G 5 g
D38951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38955G Supplier TCI America Supplier No. D38955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68528-80-3
Molecular Formula C16H20N2O8
Molecular Weight (g/mol) 368.342
MDL Number MFCD00049059
InChI Key ZWIBGKZDAWNIFC-UHFFFAOYSA-N
Synonym disuccinimidyl suberate, bis 2,5-dioxopyrrolidin-1-yl octanedioate, suberic acid bis n-hydroxysuccinimide ester, di n-succinimidyl suberate, bicl100, 2,5-pyrrolidinedione, 1,1'-1,8-dioxo-1,8-octanediyl bis oxy bis, nhs-sa, disuccinimidylsuberate, dsis, n-hydroxysuccinimide suberic acid ester
PubChem CID 100658
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) octanedioate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCCC(=O)ON2C(=O)CCC2=O

Specifications

Melting Point 171°C
Color White
Quantity 5 g
Formula Weight 368.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Di(N-succinimidyl) Suberate