missing translation for 'onlineSavingsMsg'
Learn More

Di-n-propyl disulfide, 99%

Catalog No. AAL0988122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0988122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0988122 Supplier Thermo Scientific Chemicals Supplier No. L0988122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 629-19-6 | C6H14S2 | 150.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 629-19-6
Molecular Formula C6H14S2
Molecular Weight (g/mol) 150.30
MDL Number MFCD00009378
InChI Key ALVPFGSHPUPROW-UHFFFAOYSA-N
Synonym dipropyl disulfide, propyl disulfide, disulfide, dipropyl, n-propyl disulfide, di-n-propyl disulfide, 4,5-dithiaoctane, dipropyl disulphide, 1-propyldisulfanyl propane, propyldithiopropane, 1,1'-dithiodipropane
PubChem CID 12377
ChEBI CHEBI:45758
IUPAC Name 1-(propyldisulfanyl)propane
SMILES CCCSSCCC

Specifications

Melting Point -86°C
Density 0.96
Boiling Point 193°C to 194°C
Flash Point 64°C (147°F)
Odor Unpleasant
Refractive Index 1.4967
Quantity 100 g
UN Number UN3082
Beilstein 969200
Formula Weight 150.31
Percent Purity 99%
Chemical Name or Material Di-n-propyl disulfide
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H319-H335-H227
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 211-079-8
RTECSNumber JO1955000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only