missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl Phtalate 98,0+%, TCI America™

Catalog No. P0304500ML
Change view
Click to view available options
Quantité:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
P0304500ML 500 mL
P030425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0304500ML Supplier TCI America Supplier No. P0304500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117-84-0
Formule moléculaire C24H38O4
Poids moléculaire (g/mol) 390.564
Numéro MDL MFCD00015292
Clé InChI MQIUGAXCHLFZKX-UHFFFAOYSA-N
Synonyme dioctyl phthalate, di-n-octyl phthalate, dinopol nop, n-octyl phthalate, vinicizer 85, phthalic acid, dioctyl ester, polycizer 162, dnop, phthalic acid di-n-octyl ester, dioctyl 1,2-benzenedicarboxylate
PubChem CID 8346
ChEBI CHEBI:34679
Nom de l’IUPAC Dioctylbenzène-1,2-dicarboxylate
SOURIRES CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCC

Specifications

Nom ou substance chimique Di-n-octyl Phthalate
Couleur Jaune
Point d’ébullition 380 °C
Quantité 500 mL
Poids de formule 390.56
Pourcentage de pureté ≥98,0% (GC)
Forme physique Liquide
EINECSNumber (3)-1307
RTECSNumber TI1925000
TSCA Oui