missing translation for 'onlineSavingsMsg'
Learn More

Di-n-heptyl phthalate, 98+%

Catalog No. AAA1546618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1546618 50 g
AAA1546609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1546618 Supplier Thermo Scientific Chemicals Supplier No. A1546618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3648-21-3 | C22H34O4 | 362.51 g/mol

Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-heptyl Phthalate is a Phthalate derivative and an organic extract contaminant found in drinking water, which was shown to activate Nrf-2-Mediated Antioxidant response in human cell line.

Solubility
Sparingly soluble in water.

Notes
Incompatible with Strong acids, strong bases,.

Chemical Identifiers

CAS 3648-21-3
Molecular Formula C22H34O4
Molecular Weight (g/mol) 362.51
MDL Number MFCD00043680
InChI Key JQCXWCOOWVGKMT-UHFFFAOYSA-N
Synonym diheptyl phthalate, di-n-heptyl phthalate, heptyl phthalate, di-n-heptylphthalate, phthalic acid, diheptyl ester, phthalic acid diheptyl ester, 1,2-benzenedicarboxylic acid, diheptyl ester, n-diheptyl phthalate, unii-ie05vo8p8p, diheptyl ester of phthalic acid
PubChem CID 19284
ChEBI CHEBI:34677
IUPAC Name diheptyl benzene-1,2-dicarboxylate
SMILES CCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC

Specifications

Density 0.992
Boiling Point 360°C
Flash Point 213°C (415°F)
Refractive Index 1.486
Quantity 50 g
Beilstein 2336623
Solubility Information Sparingly soluble in water.
Formula Weight 362.51
Percent Purity ≥98%
Chemical Name or Material Di-n-heptyl phthalate
Hazard Category Warning
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-885-4
RTECSNumber TI1090000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only