missing translation for 'onlineSavingsMsg'
Learn More

Di-2-pyridyl Ketone 98.0+%, TCI America™

Catalog No. D17085G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17085G Supplier TCI America Supplier No. D17085G

Chemical Identifiers

CAS 19437-26-4
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD00006288
InChI Key QPOWUYJWCJRLEE-UHFFFAOYSA-N
Synonym di-2-pyridyl ketone, di pyridin-2-yl methanone, bis 2-pyridyl ketone, 2,2'-carbonyldipyridine, di 2-pyridyl ketone, methanone, di-2-pyridinyl, 2,2'-dipyridyl ketone, 2-pyridine-2-carbonyl pyridine, pyridyl ketone
PubChem CID 88065
IUPAC Name dipyridin-2-ylmethanone
SMILES C1=CC=NC(=C1)C(=O)C2=CC=CC=N2

Specifications

Melting Point 55°C
Color Yellow
Quantity 5 g
Formula Weight 184.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-2-pyridyl Ketone