missing translation for 'onlineSavingsMsg'
Learn More

Dextrose, Monohydrate, Powder, USP, 97.5-102%, Spectrum™ Chemical

Catalog No. 18602119
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602119 12 kg Poly Pail
18602121 100 g Poly Pail
18602117 500 g Poly Pail
18602118 2.5 kg Poly Pail
18602120 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602119 Supplier Spectrum Chemical Mfg Cor Supplier No. DE14512KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DE145, 77938-63-7, C6H12O6·H2O

Spectrum™ Chemical Dextrose, Monohydrate, Powder, USP - All Spectrum Chemical USP products, including all USP dextrose products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77938-63-7
Molecular Formula C6H14O7
Molecular Weight (g/mol) 198.17
MDL Number MFCD00149450
InChI Key SPFMQWBKVUQXJV-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal hydrate
SMILES O.OCC(O)C(O)C(O)C(O)C=O

Specifications

CAS Max % 100%
Packaging Poly Pail
Quantity 12 kg
Percent Purity 97.5 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Dextrose, Monohydrate, Powder