missing translation for 'onlineSavingsMsg'
Learn More

Dextrose, Monohydrate, Powder, Spectrum™ Chemical

Catalog No. 18610151
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610151 45 kg Poly Bottle
18610149 500 g Poly Bottle
18610150 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610151 Supplier Spectrum Chemical Mfg Cor Supplier No. D101845KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77938-63-7
Molecular Formula C6H14O7
Molecular Weight (g/mol) 198.17
InChI Key SPFMQWBKVUQXJV-UHFFFAOYNA-N
IUPAC Name 2,3,4,5,6-pentahydroxyhexanal hydrate
SMILES O.OCC(O)C(O)C(O)C(O)C=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 45 kg
Grade Reagent
Insoluble Matter 5e-05
Physical Form Powder or Waxy Solid
Chemical Name or Material Dextrose, Monohydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.