missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Monohydrate, Powder, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Grade | Reagent |
| Insoluble Matter | 5e-05 |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Dextrose, Monohydrate |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.