missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Anhydrous, EP, BP, JP, USP, 99.5%, Spectrum™ Chemical
DE140, 50-99-7, C6H12O6
Supplier: Spectrum Chemical Mfg Cor DE14012KG
Description
Spectrum™ Chemical Dextrose, Anhydrous, USP, EP, BP, JP - All Spectrum Chemical USP, EP, BP, JP products, including all USP dextrose products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 50-99-7 | |
| 0.1% | |
| Poly Pail | |
| 12 kg | |
| OCC1OC(O)C(O)C(O)C1O | |
| 180.16 | |
| Multi Compendia |
| 100% | |
| 1% | |
| C6H12O6 | |
| WQZGKKKJIJFFOK-UHFFFAOYNA-N | |
| 6-(hydroxymethyl)oxane-2,3,4,5-tetrol | |
| 99.5% |
Chemical Identifiers
| 50-99-7 | |
| 180.16 | |
| 6-(hydroxymethyl)oxane-2,3,4,5-tetrol |
| C6H12O6 | |
| WQZGKKKJIJFFOK-UHFFFAOYNA-N | |
| OCC1OC(O)C(O)C(O)C1O |