missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose anhydre (granules cristallins/biologie moléculaire), Fisher BioReagents
Description
Le dextrose est souvent utilisé comme additif pour culture cellulaire.
Spécifications
Spécifications
| Nom ou substance chimique | Dextrose Anhydrous |
| Point de fusion | 146,1 °C |
| Couleur | Blanc |
| pH | 5.9 |
| Remarques sur le degré de pureté | Sans DNASE, RNase et Protéase |
| Fourchette de pourcentages d’essai | ≥99.8 % |
| Emballage | Bouteille en polyéthylène |
| Formule linéaire | HOCH2(CHOH)4CHO |
| Quantité | 1 kg |
| Indice de Merck | 15, 4495 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:1 Flammability:0 Instability:0 |
| Recommended Storage | RT |