missing translation for 'onlineSavingsMsg'
Learn More

Dextrorphan tartrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013113
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013113 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013113 Supplier Merck Emd Millipore Supplier No. PHR197430MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 143-98-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H23 NO · C4H6O6
Synonym Dextrorphan tartrate; (+)-3-Hydroxy-N-methylmorphinan (+)-tartrate salt; Dextrorphan D-tartrate
Type Certified Reference Material
Quantity 30 mg
Formula Weight 407.46
Grade Certified Reference Material