missing translation for 'onlineSavingsMsg'
Learn More

Sel de sodium sulfate de dextrane (400 000-600 000 M.Wt), MP Biomedicals™

Numéro de catalogue. ICN16011180
Change view
Click to view available options
Quantité:
1 g
10 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
ICN16011180 100 g
ICN16011101 1 g
ICN16011110 10 g
ICN16011150 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN16011180 Supplier MP Biomedicals Inc Supplier No. 0216011180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9011-18-1
Formule moléculaire (C6H7O5)m(C6H7O5)n
Poids moléculaire (g/mol) 40,000
Numéro MDL MFCD00081551
Clé InChI BLFLLBZGZJTVJG-UHFFFAOYSA-N
Synonyme benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
PubChem CID 2337
ChEBI CHEBI:116735
SOURIRES *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Specifications

Couleur Blanc
Nom ou substance chimique Dextran Sulfate Sodium Salt
Forme physique Poudre
Quantité 100 g
Nom de l’IUPAC Éthyle 4-aminobenzoate
Poids de formule 400 000 à 600 000