missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, MP Biomedicals™
Description
Depending upon its molecular weight it has different uses in different fields.
- May be used as long hydrophilic spacer arms to improve the freedom of movement of conjugated/bound proteins.
- May be derivatized for use in biosensor systems
- Other applications include: platelet aggregant, plasma volume extender, osmotic pressure regulator, stabilizer, organ separation media, matrix component, copolymer, microcarrier, binding agent, viscosity modifier, antithrombotic, lubricant and physical structure component.
Specifications
Specifications
| Physical Form | Powder |
| Solubility Information | Very soluble in water (50mg/mL), slightly soluble in alcohol. |
| Quantity | 500 g |
| Formula Weight | 60,000-90,000 |
| Grade | Clinical |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15-30°C). |