missing translation for 'onlineSavingsMsg'
Learn More

Dextran from Leuconostoc mesenteroides, For GPC, Mw 50,000, MilliporeSigma™ Supelco™

Catalog No. 111015815
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015815 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015815 Supplier Merck Emd Millipore Supplier No. 31420100MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dextran powder for use in research applications.

Food and Beverages

Chemical Identifiers

Molecular Weight (g/mol) 504.4
MDL Number MFCD00130935

Specifications

Physical Form Neat
CAS 9004-54-0
Quantity 100 mg
Linear Formula (C6H10O5)n
Grade GPC