missing translation for 'onlineSavingsMsg'
Learn More

Dextran from Leuconostoc mesenteroides, For GPC, Mw 25,000, MilliporeSigma™ Supelco™

Catalog No. 111015814
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015814 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015814 Supplier Merck Emd Millipore Supplier No. 31419100MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Food and Beverages

Chemical Identifiers

MDL Number MFCD00130935

Specifications

Physical Form Neat
CAS 9004-54-0
Linear Formula (C6H10O5)n
Quantity 100 mg
Grade GPC