missing translation for 'onlineSavingsMsg'
Learn More

Dextran, for GPC 12,000, MilliporeSigma™ Supelco™

Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

$474.90 - $1698.22

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 9004-54-0
Physical Form Powder/Crystals
Linear Formula [C6H10O5 ]n
Technique GPC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015803
View Documents
Merck Emd Millipore
00270100MG
100 mg
Each for $474.90
Only null left
 
111015821
View Documents
Merck Emd Millipore
00270500MG
500 mg
Each for $1,698.22
Only null left
 
Description

Description

Featured Industries: Food and Beverages
Specifications

Specifications

Bottomless Glass Bottle with Inserted Fused Cone
Powder/Crystals
GPC
Analytical Standard
9004-54-0
[C6H10O5 ]n
MFCD00130935
Analytical Standard
Videos
SDS
Documents

Documents