missing translation for 'onlineSavingsMsg'
Learn More

Dextran, for GPC 12,000, MilliporeSigma™ Supelco™

Catalog No. 111015803
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015803 100 mg
111015821 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015803 Supplier Merck Emd Millipore Supplier No. 00270100MG

Dextran is a polysaccharide. Dextran fluids behave as colloids; and also are used as plasma substitution. It may have siliconizing effect helping in coating raw serosal surfaces.

Featured Industries: Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 9004-54-0
Physical Form Powder/Crystals
Linear Formula [C6H10O5 ]n
Technique GPC
MDL Number MFCD00130935
Type Analytical Standard
Quantity 100 mg
Grade Analytical Standard