missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dexmedetomidine hydrochloride

Catalog No. AC465432500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465432500 250 mg
AC465430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465432500 Supplier Thermo Scientific Chemicals Supplier No. 465432500
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145108-58-3
Molecular Formula C13H17ClN2
Molecular Weight (g/mol) 236.74
InChI Key VPNGEIHDPSLNMU-MERQFXBCSA-N
IUPAC Name hydrogen 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride
SMILES [H+].[Cl-].C[C@H](C1=CN=CN1)C1=CC=CC(C)=C1C

Specifications

Melting Point 158.0°C
Quantity 250 mg
Formula Weight 236.74
Chemical Name or Material Dexmedetomidine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only