missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 99.0+%, TCI America™

Catalog No. D19611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19611G Supplier TCI America Supplier No. D19611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-02-2
Molecular Formula C22H29FO5
Molecular Weight (g/mol) 392.47
MDL Number MFCD00064136
InChI Key UREBDLICKHMUKA-CXSFZGCWSA-N
Synonym dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
PubChem CID 5743
ChEBI CHEBI:41879
IUPAC Name (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

Specifications

Melting Point 264°C
Color White
Quantity 1 g
Formula Weight 392.47
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dexamethasone
RTECSNumber TU3980000
TSCA Yes