missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014334
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014334 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014334 Supplier Merck Emd Millipore Supplier No. PHR15261G

missing translation for 'validMainframeMsg'

Dexamethasone is a corticosteroid medication, with anti-inflammatory activity. It is widely used for inflammatory ocular conditions and for the treatment of skin-allergic reactions.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 50-02-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C22H29 FO5
MDL Number MFCD00064136
Melting Point 262°C to 264°C
Synonym Dexamethasone; (11 beta,16 alpha)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione; 9 alpha-Fluoro-16 alpha-methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione; 9 alpha-Fluoro-16 alpha-methylprednisolone; Prednisolone F
Type Certified Reference Material
Quantity 1 g
Formula Weight 392.46
Grade Certified Reference Material