missing translation for 'onlineSavingsMsg'
Learn More

Dexamethasone 21-Phosphate Disodium Salt, MP Biomedicals™

Catalog No. ICN15756591
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15756591 1 g
ICN15756580 100 mg
ICN15756590 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15756591 Supplier MP Biomedicals Inc Supplier No. 0215756591

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2392-39-4
Molecular Formula C22H28FNa2O8P
Molecular Weight (g/mol) 516.41
MDL Number MFCD00079105
InChI Key PLCQGRYPOISRTQ-FCJDYXGNSA-L
Synonym dexamethasone sodium phosphate, dexamethasone 21-phosphate disodium salt, dalalone, dexadreson, dexagro, megacort, soldesam, spersadox, decdan, solu-decadron
PubChem CID 16961
ChEBI CHEBI:4462
IUPAC Name disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COP(=O)([O-])[O-])O)C)O)F)C.[Na+].[Na+]

Specifications

Quantity 1 g
Formula Weight 516.4
Physical Form Solid
Chemical Name or Material Dexamethasone 21-Phosphate Disodium Salt
Recommended Storage Store at +4°C.