missing translation for 'onlineSavingsMsg'
Learn More

Devardas alloy, MilliporeSigma™ Supelco™

Catalog No. 111018753
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018753 250 g
111018184 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018753 Supplier Merck Emd Millipore Supplier No. 31385250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Devarda's alloy is a reducing agent, and has found application in being used to reduce NO3--N, or NO2--N, into NH4+-N, thereby finding functionality in being used for total nitrogen determination.

Food and Beverages, Forensics and Toxicology

Chemical Identifiers

MDL Number MFCD00198176

Specifications

Melting Point 550°C
Quantity 250 g
CAS 8049-11-4
Form Powder
Linear Formula AlCuZn
Grade puriss. p.a.