Learn More
Thermo Scientific Chemicals Devarda's Alloy, granular
Description
Devarda's Alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline conditions. It is also used in the quantitative or qualitative analysis of nitrates in agriculture and soil science before the development of ion chromatography. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 4.355 |
| Quantity | 100 g |
| Form | Granular |
| Solubility Information | Insoluble in water. Slightly soluble in hydrochloric acid |
| Chemical Name or Material | Devarda's Alloy |
Safety and Handling
| missing translation for 'healthHazard1' | H228-H500 |
| missing translation for 'healthHazard3' | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only