missing translation for 'onlineSavingsMsg'
Learn More

Desoxyanisoin, 98%

Catalog No. AAA1281614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1281614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1281614 Supplier Thermo Scientific Chemicals Supplier No. A1281614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-44-5 | C16H16O3 | 256.301 g/mol

Deoxyanisoin react to produce a-bromo-4,4'-dimethoxy-deoxybenzoin, and this reaction could happen in the reagent of CCl4 and bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Deoxyanisoin react to produce a-bromo-4,4′-dimethoxy-deoxybenzoin, and this reaction could happen in the reagent of CCl4 and bromine.

Solubility
Sparingly soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 120-44-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00008406
InChI Key SICBLYCPRWNHHP-UHFFFAOYSA-N
Synonym desoxyanisoin, deoxyanisoin, 1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxydeoxybenzoin, deoxy-p-anisoin, deoxy-4-anisoin, 1,2-bis 4-methoxyphenyl ethan-1-one, ethanone, 1,2-bis 4-methoxyphenyl, 4-methoxybenzyl 4-methoxyphenyl ketone, acetophenone, 4'-methoxy-2-p-methoxyphenyl
PubChem CID 67120
IUPAC Name 1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 109°C to 113°C
Quantity 25 g
Beilstein 1536594
Solubility Information Sparingly soluble in water.
Formula Weight 256.3
Percent Purity 98%
Chemical Name or Material Deoxyanisoin
EINECSNumber 204-396-8
TSCA No

RUO – Research Use Only