missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Desonide

Catalog No. AC467452500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467452500 250 mg
AC467450010 1 g
AC467450050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC467452500 Supplier Thermo Scientific Chemicals Supplier No. 467452500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 638-94-8 | C24H32O6, C24H32O6 | 416.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-94-8
Molecular Formula C24H32O6

Specifications

Quantity 250 mg
Melting Point 275.0°C
Formula Weight 416.51
Chemical Name or Material Desonide
EINECSNumber 211-351-6

RUO – Research Use Only