accessibility menu, dialog, popup

UserName

CAS RN 638-94-8

IUPAC Name: (2S,4R,8S,12S)-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
Synonyms: desonide
Molecular Weight (g/mol): 416.51
Molecular Formula: C24H32O6
InChi Key: WBGKWQHBNHJJPZ-YWZQBGSISA-N
SMILES: CC1(C)O[C@@H]2C[C@H]3C4CCC5=CC(=O)C=CC5(C)[C@H]4C(O)CC3(C)[C@@]2(O1)C(=O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)