missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desogestrel Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 54024-22-5 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C22H30 O |
| MDL Number | MFCD00869346 |
| Synonym | Desogestrel; 13-Ethyl-11-methylene-18,19-dinor-17 alpha-4-pregnen-20-yn-17-ol |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 310.47 |
| Grade | Certified Reference Material |