Learn More
Thermo Scientific™ Depyrogenated Glass Containers
Save on expensive capital equipment investments with ready-to-use glassware in a wide variety of sizes and configurations.
$662.56 - $1,511.35
Specifications
| Material | Glass |
|---|---|
| Certifications/Compliance | Meet endotoxin levels of less than 0.06 EU/mL; Includes certificate of analysis |
| Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Color | No. per Case | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Color | No. per Case | Price | Quantity & Availability | ||
12200121
![]() |
Thermo Scientific™
23 CTP4/PF |
4 mL | Vial | Clear | 72 |
Case of 72 Each for $1,474.91
|
|
||
12100006
![]() |
Thermo Scientific™
139 20C/CT/LP/PF |
22 mL | Vial | Clear | 72 |
Case of 72 Each for $1,229.82
|
|
||
12100382
![]() |
Thermo Scientific™
C2002A/PF |
60 mL | Amber Packer Bottle | Amber | 24 |
Case of 12 Each for $730.65
|
|
||
12100380
![]() |
Thermo Scientific™
13002C/PF |
60 mL | Sample Jar | Clear | 24 |
Case of 24 Each for $687.53
|
|
||
12200112
![]() |
Thermo Scientific™
130 04A/PF |
125 mL | Sample Jar | Amber | 24 |
Case of 24 Each for $1,511.35
|
|
||
12200113
![]() |
Thermo Scientific™
130 04C/TL/PF |
125 mL | Sample Jar | Clear | 24 |
Case of 24 Each for $719.29
|
|
||
12100383
![]() |
Thermo Scientific™
C2004A/PF |
125 mL | Packer Bottle | Amber | 24 |
Case of 24 Each for $719.29
|
|
||
12100004
![]() |
Thermo Scientific™
131 08A/PF |
250 mL | Sample Jar | Amber | 24 |
Case of 24 Each for $758.79
|
|
||
12200105
![]() |
Thermo Scientific™
12108A/PF |
250 mL | Packer Bottle | Amber | 12 |
Case of 12 Each for $662.56
|
|
||
12200114
![]() |
Thermo Scientific™
131 08C/PF |
250 mL | Sample Jar | Clear | 24 |
Case of 24 Each for $737.44
|
|
||
12200107
![]() |
Thermo Scientific™
122 16A/PF |
500 mL | Packer Bottle | Amber | 12 |
Case of 12 Each for $1,154.03
|
|
||
12200158
![]() |
Thermo Scientific™
132 16C/PF |
500 mL | Sample Jar | Clear | 12 |
Case of 12 Each for $764.68
|
|
||
12200115
![]() |
Thermo Scientific™
133 32C/PF |
1000 mL | Wide Mouth Bottle | Clear | 12 |
Case of 12 Each for $717.03
|
|
||
12200108
![]() |
Thermo Scientific™
123 32A/PF |
1000 mL | Container | Amber | 12 |
Case of 12 Each for $764.68
|
|
||
12200109
![]() |
Thermo Scientific™
123 40A/PF |
1200 mL | Packer Bottle | Amber | 24 |
Case of 24 Each for $1,089.15
|
|
||
12100379
![]() |
Thermo Scientific™
1172L/PF |
2000 mL | Wide Mouth Bottle | Clear | 6 |
Case of 6 Each for $976.41
|
|
||
12200110
![]() |
Thermo Scientific™
123 80A/PF |
2500 mL | Packer Bottle | Amber | 4 |
Case of 4 Each for $743.59
|
|
||
Description
Save on expensive capital equipment investments with ready-to-use glassware in a wide variety of sizes and configurations. Thermo Scientific™ Depyrogenated Glass Containers are specially prepared to meet endotoxin levels of less than 0.06 EU/mL for use in terminal sterilization, water sampling and product storage.
- Cleaning and packaging is performed in a Class 100/10 Cleanroom
- Certificate of Analysis (COA) is included with each lot
Recommended for:
Endotoxin-free applications
Ordering Information:
Containers with additional low particle certification are available. Thermo Scientific also offers glass vials specially prepared to meet endotoxin levels of less than 0.06 EU/mL. Ask your customer service representative about our depyrogenation service with our validated process, performed in our Class 100 Cleanroom.
Specifications
| Glass | |
| Meet endotoxin levels of less than 0.06 EU/mL; Includes certificate of analysis |