missing translation for 'onlineSavingsMsg'
Learn More

Density Standard 1191 kg/m3 HD Fitzgerald Ltd. Quality, MilliporeSigma™ Supelco™

Catalog No. 111018440
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018440 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018440 Supplier Merck Emd Millipore Supplier No. 8935310ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 131-11-3
Ignition Point 1033°F
Density 1.19 g/mL (at 25°C)
Flash Point 146.0°C - closed cup
Linear Formula C6H4-1,2-(CO2 CH3)2
MDL Number MFCD00008425
Melting Point 2°C
Boiling Point 282°C
Synonym Dimethyl phthalate; DMP
Type Organic Standard
Quantity 10 mL
Formula Weight 194.18
Grade H&D; Fitzgerald Ltd. Quality
Show More Show Less