missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Metal Density Cube Without Hook

Catalog No. S24860
Change view
Click to view available options
Material:
Aluminum
Brass, Lead, Zinc, Copper, Aluminum, Iron
Copper
Tin
Quantity:
1/Ea.
6/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Quantity
S24860 Brass, Lead, Zinc, Copper, Aluminum, Iron 6/Pk.
S27640 Copper 1/Ea.
S12915 Aluminum 1/Ea.
S27352 Tin 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24860 Supplier Eisco™ Supplier No. PH0108H

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for density exploration and experiments

  • Solid cubes, 20 mm each size
  • Stamped with element

Specifications

Quantity 6/Pk.
For Use With (Application) Density, Specific Gravity, Specific Heat Activities
Product Type Density Cube
Length (Metric) 20 mm
Length (English) 0.8 in.
Includes Six 20 mm cubes of the following metals: brass, lead, zinc, copper, aluminum, and iron
Material Brass, Lead, Zinc, Copper, Aluminum, Iron